Learn More
Electron Microscopy Sciences EMS High Alloy DX Biological Tweezers Style 7, 115 mm OAL, 0.07 mm Tip width
SDP
Catalog No. 501930613
Encompass_Preferred
The high alloy biological tweezers are anti-acid and anti-magnetic. The handles are serrated. Tips: Very fine, curved, superior finish.
OAL: 115 mm
Tip width: 0.07 mm
Tip thickness: 0.03 mm
Catalog No. 50-193-0613 Supplier Electron Microscopy Sciences Supplier No. 783257DX
The high alloy biological tweezers are anti-acid and anti-magnetic. The handles are serrated. Tips: Very fine, curved, superior finish.
OAL: 115 mm
Tip width: 0.07 mm
Tip thickness: 0.03 mm