Learn More
TrippNT DX CART SILVER W/ DOOR
SDP
Supplier:  TrippNT 53519
Encompass_Preferred
TrippNT's Core DX Cart with Metallic Silver Drawers and Hinged Door organizes, stores, and secures supplies. It offers 3 total locking drawers, 4 shelves, 2 dispensing shelves, and bulk storage with a hinged clear door. Its built-in handles are ergonomic and easy to grab. The Hinged door allows simultaneous access to the bulk and drawer storage. It is constructed from a corrosion-resistant non-metal polymer with robust 4-inch twin wheel casters capable of easily navigating elevator gaps and uneven surfaces. Available in 6 color variants. Ships fully assembled.
Catalog No. NC3602823