Learn More
Optimize Technologies Deuterium Lamp, Agilent/HP C Series, 1050 C, 1050 DAD, 1090, 1040, 79583C
SDP
Catalog No. 50113752
Encompass
Optimize offers a broad selection of lamps for OEM HPLC detectors. The high quality standard lamps come with a guaranteed 1,000 hour lifetime. We also offer our DX2 DoubleLife» Lamps. These long-life lamps are made using only top quality components and come with a 2,000 hour lifetime guarantee. This item is equivalent to OEM part number 79883-60002.
Catalog No. 50-113-752 Supplier Optimize Technologies Supplier No. 273202844
Optimize offers a broad selection of lamps for OEM HPLC detectors. The high quality standard lamps come with a guaranteed 1,000 hour lifetime. We also offer our DX2 DoubleLife» Lamps. These long-life lamps are made using only top quality components and come with a 2,000 hour lifetime guarantee. This item is equivalent to OEM part number 79883-60002.