Learn More
Optimize Technologies Deuterium DoubleLife Lamp, Waters 996, 2996, PDA
SDP
Catalog No. 50114174
Encompass
Optimize offers a broad selection of lamps for OEM HPLC detectors. The high quality standard lamps come with a guaranteed 1,000 hour lifetime. We also offer our DX2 DoubleLife» Lamps. These long-life lamps are made using only top quality components and come with a 2,000 hour lifetime guarantee. This item is designed to work with Waters 996, 2996, and PDA pump models and is equivalent to OEM part number WAT052586.
Catalog No. 50-114-174 Supplier Optimize Technologies Supplier No. 393202835
Optimize offers a broad selection of lamps for OEM HPLC detectors. The high quality standard lamps come with a guaranteed 1,000 hour lifetime. We also offer our DX2 DoubleLife» Lamps. These long-life lamps are made using only top quality components and come with a 2,000 hour lifetime guarantee. This item is designed to work with Waters 996, 2996, and PDA pump models and is equivalent to OEM part number WAT052586.