Learn More
Optimize Technologies Deuterium DoubleLife Lamp, Agilent/HP 1100, 1200 DAD
SDP
Catalog No. 50113753
Encompass
Optimize offers a broad selection of lamps for OEM HPLC detectors. The high quality standard lamps come with a guaranteed 1,000 hour lifetime. We also offer our DX2 DoubleLife» Lamps. These long-life lamps are made using only top quality components and come with a 2,000 hour lifetime guarantee. This item is equivalent to OEM part number 5182-1530.
Catalog No. 50-113-753 Supplier Optimize Technologies Supplier No. 273202845
Optimize offers a broad selection of lamps for OEM HPLC detectors. The high quality standard lamps come with a guaranteed 1,000 hour lifetime. We also offer our DX2 DoubleLife» Lamps. These long-life lamps are made using only top quality components and come with a 2,000 hour lifetime guarantee. This item is equivalent to OEM part number 5182-1530.