Learn More
Optimize Technologies Deuterium DoubleLife Lamp, Shimadzu SPD-10A, -10AV, -M10A, -10AS, -10AVP, -M10AVP, -20AV
SDP
Catalog No. 50113942
Encompass
Optimize offers a broad selection of lamps for OEM HPLC detectors. The high quality standard lamps come with a guaranteed 1,000 hour lifetime. We also offer our DX2 DoubleLife» Lamps. These long-life lamps are made using only top quality components and come with a 2,000 hour lifetime guarantee. This item is equivalent to OEM part number 228-34016-02.
Catalog No. 50-113-942 Supplier Optimize Technologies Supplier No. 343202837
Optimize offers a broad selection of lamps for OEM HPLC detectors. The high quality standard lamps come with a guaranteed 1,000 hour lifetime. We also offer our DX2 DoubleLife» Lamps. These long-life lamps are made using only top quality components and come with a 2,000 hour lifetime guarantee. This item is equivalent to OEM part number 228-34016-02.