Learn More
Medchemexpress LLC Dx3-213b | 2749555-66-4 | 99.7% | 478.57 g/mol | C20H28F2N2O5S2 | 5 MG
SDP
Catalog No. 5000221440
Encompass_Preferred
DX3-213B is a potent, orally active oxidative phosphorylation (OXPHOS) complex I inhibitor used in preclinical research. It disrupts mitochondrial ATP production and exhibits cellular activity in cancer models, making it useful for mechanistic studies of mitochondrial function and cancer metabolism. Not for human use.
  • Highly potent complex I inhibition (IC50 = 3.6 nM)
  • Impairs ATP generation in cells (IC50 = 11 nM)
  • Inhibits growth of pancreatic cancer cell line (GI50 = 11 nM)
  • High chemical purity suitable for biochemical assays
  • Available as solid and DMSO solution for assay flexibility
  • Intended for mechanistic mitochondrial and cancer metabolism research
Catalog No. 50-002-21440 Supplier Medchemexpress LLC Supplier No. HY1443105MG
DX3-213B is a potent, orally active oxidative phosphorylation (OXPHOS) complex I inhibitor used in preclinical research. It disrupts mitochondrial ATP production and exhibits cellular activity in cancer models, making it useful for mechanistic studies of mitochondrial function and cancer metabolism. Not for human use.
  • Highly potent complex I inhibition (IC50 = 3.6 nM)
  • Impairs ATP generation in cells (IC50 = 11 nM)
  • Inhibits growth of pancreatic cancer cell line (GI50 = 11 nM)
  • High chemical purity suitable for biochemical assays
  • Available as solid and DMSO solution for assay flexibility
  • Intended for mechanistic mitochondrial and cancer metabolism research