Learn More
Medchemexpress LLC DX3-213B | 2749555-66-4 | 99.7% | C20H28F2N2O5S2 | 100 MG
SDP
Catalog No. 5000355093
Encompass_Preferred
DX3-213B is a highly potent, orally active oxidative phosphorylation (OXPHOS) complex I inhibitor with an IC50 of 3.6 nM. It effectively impairs ATP generation with an IC50 of 11 nM and blocks the growth of MIA PaCa-2 cells, also with a GI50 of 11 nM. This compound is utilized in research concerning pancreatic cancer.
  • Highly potent and orally active.
  • Functions as an oxidative phosphorylation (OXPHOS) complex I inhibitor.
  • Impairs ATP generation.
  • Blocks MIA PaCa-2 cell growth.
  • Used in pancreatic cancer research.
Catalog No. 50-003-55093 Supplier Medchemexpress LLC Supplier No. HY144310100MG
DX3-213B is a highly potent, orally active oxidative phosphorylation (OXPHOS) complex I inhibitor with an IC50 of 3.6 nM. It effectively impairs ATP generation with an IC50 of 11 nM and blocks the growth of MIA PaCa-2 cells, also with a GI50 of 11 nM. This compound is utilized in research concerning pancreatic cancer.
  • Highly potent and orally active.
  • Functions as an oxidative phosphorylation (OXPHOS) complex I inhibitor.
  • Impairs ATP generation.
  • Blocks MIA PaCa-2 cell growth.
  • Used in pancreatic cancer research.