Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DX600 | 478188-26-0 | 99.7% | 3074.33 | 5 MG
SDP

Catalog No. 5000423003
Encompass_Preferred

DX600 is a selective ACE2 specific inhibitor (KD: 1.3 nM) that does not cross-react with ACE. It has been shown to exacerbate diabetes-induced cardiovascular dysfunction and increase cardiac and renal NOX activity.

  • Selective ACE2 specific inhibitor (KD: 1.3 nM)
  • Does not cross-react with ACE
  • Inhibits rhACE2 activity by 47% at 1 μM
  • Inhibits ACE2 activity by 42% in human MNCs at 10 μM
  • Decreases NR 8383 cell growth and increases TNF-a and IL-6 content at 100 nM
  • Exacerbates diabetes-induced cardiovascular dysfunction in diabetic rats
  • Increases thrombus weight by 30% in a thrombosis model in rats

Catalog No. 50-004-23003 Supplier Medchemexpress LLC Supplier No. HYP22225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DX600 is a selective ACE2 specific inhibitor (KD: 1.3 nM) that does not cross-react with ACE. It has been shown to exacerbate diabetes-induced cardiovascular dysfunction and increase cardiac and renal NOX activity.

  • Selective ACE2 specific inhibitor (KD: 1.3 nM)
  • Does not cross-react with ACE
  • Inhibits rhACE2 activity by 47% at 1 μM
  • Inhibits ACE2 activity by 42% in human MNCs at 10 μM
  • Decreases NR 8383 cell growth and increases TNF-a and IL-6 content at 100 nM
  • Exacerbates diabetes-induced cardiovascular dysfunction in diabetic rats
  • Increases thrombus weight by 30% in a thrombosis model in rats

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.