Learn More
Medchemexpress LLC DX600 TFA | 99.2% | 3074.33 (free acid) | 1 MG
SDP
Catalog No. 5000422810
Encompass_Preferred
DX600 TFA is a selective ACE2 specific inhibitor (KD: 1.3 nM) that does not cross-react with ACE. It has been observed to exacerbate diabetes-induced cardiovascular dysfunction and increase cardiac and renal NOX activity. It is for research use only.
  • Selective ACE2 specific inhibitor
  • Does not cross-react with ACE
  • Exacerbates diabetes-induced cardiovascular dysfunction
  • Increases cardiac and renal NOX activity
Catalog No. 50-004-22810 Supplier Medchemexpress LLC Supplier No. HYP2222A1MG
DX600 TFA is a selective ACE2 specific inhibitor (KD: 1.3 nM) that does not cross-react with ACE. It has been observed to exacerbate diabetes-induced cardiovascular dysfunction and increase cardiac and renal NOX activity. It is for research use only.
  • Selective ACE2 specific inhibitor
  • Does not cross-react with ACE
  • Exacerbates diabetes-induced cardiovascular dysfunction
  • Increases cardiac and renal NOX activity