Learn More
Medchemexpress LLC DX600 (TFA) | 99.2% | 3074.33 (free acid) | 5 MG
SDP
Catalog No. 5000422286
Encompass_Preferred
DX600 TFA is a selective ACE2 specific inhibitor with a KD of 1.3 nM, and it does not cross-react with ACE. This compound exacerbates diabetes-induced cardiovascular dysfunction and increases cardiac and renal NOX activity. It is intended for research use only.
  • Inhibits rhACE2 activity.
  • Decreases NR 8383 cell growth and increases TNF-a and IL-6 content.
  • Exacerbates diabetes-induced cardiovascular dysfunction.
  • Increases thrombus weight in a thrombosis model.
  • Increases cardiac and renal NOX activity.
Catalog No. 50-004-22286 Supplier Medchemexpress LLC Supplier No. HYP2222A5MG
DX600 TFA is a selective ACE2 specific inhibitor with a KD of 1.3 nM, and it does not cross-react with ACE. This compound exacerbates diabetes-induced cardiovascular dysfunction and increases cardiac and renal NOX activity. It is intended for research use only.
  • Inhibits rhACE2 activity.
  • Decreases NR 8383 cell growth and increases TNF-a and IL-6 content.
  • Exacerbates diabetes-induced cardiovascular dysfunction.
  • Increases thrombus weight in a thrombosis model.
  • Increases cardiac and renal NOX activity.