Learn More
Beckman Coulter™ DXA 5000 Fit
Catalog No. 01339728 Shop All Beckman Coulter Products
Change view
Click to view available options
Includes:
1 CENTR, 1 DxI, 2 DxC 700 AU
1 CENTR, 2 DxI, 2 DxC 700 AU
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
01-339-728 1 CENTR, 2 DxI, 2 DxC 700 AU
01-339-727 1 CENTR, 1 DxI, 2 DxC 700 AU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-339-728 Supplier Beckman Coulter™ Supplier No. C70420
Add to Cart
Add to Cart
An automation solution that fundamentally improves lab workflow by making intelligent automation accessible to fit your small- and mid-sized lab needs.
The DxA 5000 Fit automation system delivers a unique combination of rapid and consistent turnaround time along with the industry's most comprehensive pre-analytical sample quality assessment. It delivers significant efficiency gains by joining automated pre- and post-analytical sample processing with connected hematology, chemistry and immunoassay analyzers.
TRUSTED_SUSTAINABILITY
Specifications
Type Automated Enzyme Immunoassay Analyzer
Throughput 4,000 Tests/Day
Includes 1 CENTR, 2 DxI, 2 DxC 700 AU