Learn More
Description
- Dynamic sample handler enables continuous testing with a single rack input/output with 12 lanes for calibration, qc, routine, or STAT patient testing
- DxLAB racks accommodate various tube and cup sizes for continuous loading so there's no need to sort by size.
- Automated HbA1c Advanced sample prep
- Re-designed intuitive user interface
- SimpleSolve Onboard Guide gives a step-by-step guide for troubleshooting
- DxS IntelliServe solution remotely monitors issues to maximize uptime
- 400 photometric tests per hour, up to 800 with ISE
- ISE maximum test per hour: 200 if ISE only
- HbA1c maximum tests per hour: 65 if HbA1c only
Specifications
Specifications
| Type | Clinical Chemistry Analyzer |
| Sample Type | Serum, Plasma, Urine, Whole Blood |
| For Use With (Equipment) | Beckman Coulter Analyzer |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.