Learn More
Description
The DxC 500i's broad and growing portfolio spans chemistry and immunoassay, and includes high-quality Six Sigma performance assays, giving you a comprehensive view of disease states with one, integrated analyzer.
Specifications
Specifications
| Type | Clinical Chemistry Analyzer |
| Sample Type | Serum, Plasma, Urine, CSF, Amniotic Fluid, Whole Blood |
| For Use With (Equipment) | Beckman Coulter Analyzers |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.