Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter™ DxC 700 AU Chemistry Analyzer

Catalog No. 22000000 Shop All Beckman Coulter Products
Change view
Click to view available options
Reagent Capacity:
350 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Reagent Capacity
22-000-000 350 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-000-000 Supplier Beckman Coulter™ Supplier No. B98654

This product is not orderable online. For ordering, please contact your Fisher Healthcare Sales Representative.

This item is not returnable. View return policy

The DxC 700 AU clinical chemistry analyzer is the latest innovation in a line of reliable chemistry systems for the mid- to high-volume laboratory. This powerful combination delivers maximal uptime, high reliability and precise performance.

  • Newly designed intuitive graphical user interface
  • Simplified reagent inventory, calibration and quality control procedures
  • Online maintenance documentation
  • User-customized menu and free-text notepad
  • Color alerts to highlight system operating conditions
  • Reliability for greater uptime with quick and easy replacement of common parts
  • No special tools required
  • No more than three steps and no longer than 60 seconds for replacement of parts, such as sample and reagent probes, mixers, syringes, ISE electrodes and optical lamp
  • Extensive online help with instructional videos
  • Available integration with pre-analytical automation through direct track sampling from Power Express
  • Available connection to Command Central with REMISOL Advance, allowing access to multiple instruments and automation consoles from a single workstation, optimizing laboratory management and improving decision making
  • PROService remote diagnostic capabilities ensure 24/7 continuous monitoring of vital system functions
  • Cooled STAT compartment, providing one-button STAT interrupt, advanced auto QC and calibration capabilities
  • Ability to load reagents while the analyzer is in measure mode
  • Priority sample repeat/reflex
  • Economical ISEs with long onboard stability; easy to maintain (only individual electrode replacement required)
  • 150-sample continuous rack loader
TRUSTED_SUSTAINABILITY

Specifications

Type Clinical Chemistry Analyzer
Sample Type Serum, Plasma, Urine, Whole Blood
Analytical Method Colorimetry, Turbidimetry, Latex Agglutination, Indirect ISE
Capacity (Metric) 350 μL
Reagent Capacity 350 μL
Cuvettes Yes
Depth (Metric) 890 mm (Analyzer), 1040 mm (Sampler)
Height (Metric) 1300 mm (Analyzer), 940 mm (Sampler)
Width (Metric) 1250 mm (Analyzer), 730 mm (Sampler)
Voltage 200/240 V
Measurement Method Arsenazo, Enzymatic
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.