Learn More
Medchemexpress LLC HY-13631D 100mg , Dxd CAS:1599440-33-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13631D100MG
Encompass_Preferred
HY-13631D 100mg Dxd CAS: 1599440-33-1 Dxd (Exatecan derivative for ADC) is a potent DNA topoisomerase I inhibitor, with an IC50 of 0.31 μM, used as a conjugated drug of HER2-targeting ADC (DS-8201a). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9502