Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dxd-d5 (exatecan-d5 derivative for ADC) | 00-00-0 | 98.9% | C26H19D5FN3O6 | 10MG
SDP

Catalog No. 5000260297
Encompass_Preferred

Dxd-d5 is a deuterium-labeled form of DXd, an exatecan-derived ADC payload and potent DNA topoisomerase I inhibitor. DXd has a reported IC50 of 0.31 μM and is used as the cytotoxic component in HER2-targeting antibody-drug conjugates. Dxd-d5 is provided for research use as a tracer and as an internal standard for quantitative analytical methods and ADC development studies.

  • Deuterium-labeled tracer suitable for metabolic and mechanistic studies.
  • Potent topoisomerase I inhibitor with reported IC50 0.31 μM.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS quantitation.
  • High reported purity appropriate for analytical applications.
  • Molecular formula C26H19D5FN3O6 and molecular weight 498.51.
  • Provided in small-scale quantities for analytical and research workflows.

Catalog No. 50-002-60297 Supplier Medchemexpress LLC Supplier No. HY13631DS10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dxd-d5 is a deuterium-labeled form of DXd, an exatecan-derived ADC payload and potent DNA topoisomerase I inhibitor. DXd has a reported IC50 of 0.31 μM and is used as the cytotoxic component in HER2-targeting antibody-drug conjugates. Dxd-d5 is provided for research use as a tracer and as an internal standard for quantitative analytical methods and ADC development studies.

  • Deuterium-labeled tracer suitable for metabolic and mechanistic studies.
  • Potent topoisomerase I inhibitor with reported IC50 0.31 μM.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS quantitation.
  • High reported purity appropriate for analytical applications.
  • Molecular formula C26H19D5FN3O6 and molecular weight 498.51.
  • Provided in small-scale quantities for analytical and research workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.