Learn More
Medchemexpress LLC Dxd-d5 (Exatecan-d5 derivative for ADC) | 1599440-33-1 | 98.9% | 498.51 | 1 ML
SDP
Catalog No. 5000349790
Encompass_Preferred
Dxd-d5 is a deuterium labeled Dxd, which is a potent DNA topoisomerase I inhibitor with an IC50 of 0.31 μM. It is used as a conjugated drug of HER2-targeting ADC (DS-8201a).
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
  • Intended for research use only.
Catalog No. 50-003-49790 Supplier Medchemexpress LLC Supplier No. HY13631DS10MM/1ML
Dxd-d5 is a deuterium labeled Dxd, which is a potent DNA topoisomerase I inhibitor with an IC50 of 0.31 μM. It is used as a conjugated drug of HER2-targeting ADC (DS-8201a).
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
  • Intended for research use only.