Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter DxH 500

Catalog No. 21403581 Shop All Beckman Coulter Products
Change view
Click to view available options
Throughput:
60 Samples/hr.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Throughput
21-403-581 60 Samples/hr.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-403-581 Supplier Beckman Coulter Supplier No. B40601

This product is not orderable online. For ordering, please contact your Fisher Healthcare Sales Representative.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The DxH 500 hematology analyzer can help you improve clinical decision-making and efficiency.

Benefit from accurate and comprehensive analysis in your low-volume lab setting. The DxH 500 hematology analyzer can help you improve clinical decision-making and efficiency. Built on proven cellular analysis technology, the DxH 500 provides precise platelet counts and accurate first-pass results. This means you can reduce costly test reruns and time-consuming manual reviews. The system's intuitive interface also makes it easier to learn and operate, freeing your staff to focus on patient care.

  • Spend time with your patients, not your analyzer
  • Maximize uptime with built-in reliability, automatic start-up, efficient reagent usage and easy reagent replacement
  • Free up time to focus on higher value activities with the ability to change all three reagents in less than five minutes
  • Use 50% less reagent, consume reagents half as fast, and take less time changing and restocking reagents with the analyzer's efficient approach
  • Leverage single-sample precision
  • Obtain accurate results with samples as small as 12 μL of blood
  • Save time by repeating testing on the same sample without another draw
  • Improve laboratory efficiency
  • Enable fast analysis with the analyzer's unique fluidic design
  • Conserve valuable bench space with the DxH 500 hematology analyzer's small footprint and integrated computer
  • Reduce operational complexity with the simplified user interface, which enables any command to be performed in three touches or less
  • Advance your efforts to go paperless with the analyzer's bi-directional LIS Interface
  • Reduce operating costs
  • Reduce disposal costs with cyanide-, azide- and formaldehyde-free reagents
  • Lower electricity bills with the DxH 500's efficient power handling
TRUSTED_SUSTAINABILITY

Specifications

Sample Type Whole Blood
For Use With (Equipment) Beckman Coulter Analyzers
Sample Size 12 μL of Veinous or Micro-collected Whole Blood, 20 μL of Whole Blood for Pre-dilute Analysis
Throughput 60 Samples/hr.
Depth (English) 17 in.
Height (English) 16 in.
Width (English) 11 in.
Weight (English) 25.1 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.