Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter DxH 500 Diluent

Catalog No. 21280101 Shop All Beckman Coulter Products
Change view
Click to view available options
Quantity:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-280-101 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-280-101 Supplier Beckman Coulter Supplier No. B36845
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

DxH 500 Series Diluent is non-toxic and can help reduce overall operating costs.

  • Reduce reagent usage - Labs use 50% less reagent volume per sample, compared to other low-volume analyzers. A single reagent bottle can support hundreds of tests.
  • Lower the cost of disposal. Cyanide-, azide-, and formaldehyde-free reagents reduce the cost of disposal as well as helping to meet environmental and regulatory compliance standards.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Beckman Coulter Analyzer
Type Diluent
Quantity 10 L
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.