Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter DxH 520 Hematology Instrument

Catalog No. 21403582 Shop All Beckman Coulter Products
Change view
Click to view available options
Throughput:
60 Samples/hr.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Throughput
21-403-582 60 Samples/hr.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-403-582 Supplier Beckman Coulter Supplier No. B40602
Request a Quote

This product is not orderable online. For ordering, please contact your Fisher Healthcare Sales Representative.

This item is not returnable. View return policy

The DxH 520 is a 5-part differential closed tube hematology analyzer, designed to help your low-volume laboratory enhance patient care through increased productivity with reduced costs.

The DxH 520 is a 5-part differential closed tube hematology analyzer, designed to help your low-volume laboratory enhance patient care through increased productivity with reduced costs. The system delivers critical results the first time with accuracy, with as little as 375 μL tube fill-volume and an aspiration of only 17 μL - ideal for pediatric or difficult-draw samples. Technologists can experience safety without compromise with closed tube technology, providing optimal security against blood-borne pathogens. The DxH 520 hematology analyzer also has one of the smallest footprints in its class, uses only three reagents, and features a robust patient data and QC management package. Automated operational tasks promote ease of use, efficiency and uptime, freeing more time for patient care.

  • Maximize laboratory productivity
  • Optimize uptime with proven system reliability and an automated cleaning cycle
  • Experience infrequent and simple reagent reloading - easily change all three reagents in less than five minutes
  • Gain powerful patient and QC data management tools for paperless operations and to reduce clerical errors
  • Discover a more efficient use of laboratory counter space with the analyzer's compact footprint - smallest in its class
  • Deliver high-quality clinical results
  • Ensure a more accurate differential with proprietary dynamic gating
  • Improve flagging and reduce slide reviews while maintaining effective clinical sensitivity
  • Reduce overall operating costs
  • Lower overhead costs by consuming fewer reagents and using less power due to a reduction in moving parts and a more intelligent, energy-optimizing design
  • Eliminate disposal costs with cyanide-, azide- and formaldehyde-free reagents
TRUSTED_SUSTAINABILITY

Specifications

Sample Type Whole Blood
For Use With (Equipment) Beckman Coulter Analyzers
Sample Size 17 μL of Veinous or Micro-collected Whole Blood, 20 μL of Whole Blood for Pre-dilute Analysis
Throughput 60 Samples/hr.
Depth (English) 16.9 in.
Height (English) 16.0 in.
Width (English) 10.6 in.
Weight (English) 25.1 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.