Description
- The DxH 900 hematology analyzer delivers accurate and high-confidence results on the first run.
- With its best-in-class hematology technology, this automated analyzer minimizes slide reviews, maximizes laboratory staff time, and provides significant cost savings.
- The DxH 900 offers scalable, customized solutions to address the hematology testing needs of high-volume labs.
- Customizable data management provides greater flexibility more actionable results
- Reduced labor and material costs through fewer confirmatory and repeat tests
- Lower cost of ownership with wide range of susceptibility tests on one panel
- Streamlined workflow supports testing for most routine samples through a more efficient Prompt™ system
Specifications
Specifications
| For Use With (Equipment) | Beckman Coulter Analyzers |
| Product Line | DxH 900 |
| Sample Size | 50 μL |
| Throughput | 100 Samples/hr. |
| Includes | UPS |