Learn More
Beckman Coulter™ DXI 800 SLGP 2
Catalog No. 21200497 Shop All Beckman Coulter Products
Change view
Click to view available options
Throughput:
400 Tests/hr.
Catalog No. Throughput
21-200-497 400 Tests/hr.
1 options
Catalog No. 21-200-497 Supplier Beckman Coulter™ Supplier No. D00185
Add to Cart
Add to Cart
Accelerate immunoassay time-to-results in your high-volume lab with the UniCel DxI 800 Access Immunoassay System.
The DxI 800 is simple enough for technologists of all skill levels to use, with industry-leading intuitive user interfaces and 'load-and-go' simplicity that frees up staff for higher value work - no pausing or standby time required.
Built for scalability
  • All of our immunoassay systems feature standardized test menus, assay protocols, instrument processes and reference ranges. Together, these features minimize training requirements and streamline inventory management tasks for labor cost savings, while providing consistent results across the healthcare delivery network
  • Results correlate across platforms for consistency in patient care
  • Continuous loading of STAT and routine samples address peak workflow requirements
Highly flexible
  • System releases the primary tube after creating an aliquot, making the system available for other uses
  • Refrigerated compartment holds 50 reagent packs in any configuration, allowing to customize the system menu to match your testing needs
  • Performs up to 400 tests per hour
  • Broad assay test menu and simple operation give you the ability to expand in-house services
  • Capable of integrating with automation and IT systems
  • Optimized for the needs of the technologists
  • Load reagents and consumables without pausing the instrument
  • Load samples and walk away to perform higher value work and return to results
  • Maximize walk-away time with large onboard supply capacity
  • View monitor and indicator lights to see which supplies need replenishment
  • Remotely diagnose analyzer with optional PROService
TRUSTED_SUSTAINABILITY
Specifications
Type Automated Enzyme Immunoassay Analyzer
Throughput 400 Tests/hr.