Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter DxM 1040 MicroScan WalkAway ID/AST System

Catalog No. 21403587 Shop All Beckman Coulter Products
Change view
Click to view available options
Includes:
LabPro Low Voltage
UPS
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
21-403-587 UPS
21-403-585 LabPro Low Voltage
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-403-587 Supplier Beckman Coulter Supplier No. C13844
Add to Cart
Add to Cart

Enable your laboratory to optimize patient care while reducing risks, costs and operational burden associated with detecting emerging antimicrobial resistance.

  • Reduced turnaround time for rapid and specialty ID panels.
  • Proven solutions through scientific expertise and proprietary antibiotic formulations.
  • Reduced offline testing provides faster turnaround time.
  • Customizable data management provides greater flexibility more actionable results.
  • Reduced labor and material costs through fewer confirmatory and repeat tests.
  • Lower cost of ownership with wide range of susceptibility tests on one panel.
  • Streamlined workflow supports testing for most routine samples through a more efficient Prompt™ system.
  • Simultaneous processing of conventional, rapid and specialty panels on a single, automated platform.
  • Easy-to-view external LED indicators show status at a glance.
  • Simplified maintenance with updated reagent-handling system.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Beckman Coulter Analyzers
Includes UPS
Depth (English) 34 in.
Height (English) 29 in.
Width (English) 38.5 in.
Weight (English) 376 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.