Learn More
Cayman Chemical DXR InhIbItr 11afree acId 1mg
SDP
Catalog No. 502942128
Encompass_Preferred
An inhibitor of P. falciparum DXR (IC50 = 0.11 µM); inhibits the growth of P. falciparum in isolated human erythrocytes (IC50 = 0.37 µM)
Catalog No. 50-294-2128 Supplier Cayman Chemical Supplier No. 397811 MG
An inhibitor of P. falciparum DXR (IC50 = 0.11 µM); inhibits the growth of P. falciparum in isolated human erythrocytes (IC50 = 0.37 µM)