Learn More
Cayman Chemical DXR Inhibitor 11a (free acid), Purity: ≥98%, CAS: 2260608-07-7, 5 mg
SDP
Catalog No. 502942106
Encompass_Preferred
An inhibitor of P. falciparum DXR (IC50 = 0.11 µM); inhibits the growth of P. falciparum in isolated human erythrocytes (IC50 = 0.37 µM)
Catalog No. 50-294-2106 Supplier Cayman Chemical Supplier No. 397815 MG
An inhibitor of P. falciparum DXR (IC50 = 0.11 µM); inhibits the growth of P. falciparum in isolated human erythrocytes (IC50 = 0.37 µM)