Learn More
Cayman Chemical DXR Inhibitor 11a (sodium salt), Purity: ≥98%, 1 mg
SDP
Catalog No. 502917151
Encompass_Preferred
An inhibitor of P. falciparum DXR (IC50 = 0.11 µM); inhibits the growth of P. falciparum in isolated human erythrocytes (IC50 = 0.37 µM)
Catalog No. 50-291-7151 Supplier Cayman Chemical Supplier No. 393051 MG
An inhibitor of P. falciparum DXR (IC50 = 0.11 µM); inhibits the growth of P. falciparum in isolated human erythrocytes (IC50 = 0.37 µM)