Learn More
Beckman Coulter™ DXU 840M Iris
Catalog No. 01361033 Shop All Beckman Coulter Products
Change view
Click to view available options
DXU 840M Iris
Catalog No.
01-361-033 DXU 840M Iris
1 options
Catalog No. 01-361-033 Supplier Beckman Coulter™ Supplier No. C81581
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Take the interruptions out of your urinalysis workflow.
Take the Interruptions Out of Your Urinalysis Workflow
The DxU Iris Workcell pairs the DxU 840/850 Iris urine microscopy analyzer with the AUTION MAX™ 4030 urine chemistry analyzer to create a scalable, fully automated urinalysis solution for your laboratory. Using proprietary Digital Flow Morphology technology with Auto-Particle Recognition (APR) software, the DxU Iris Workcell helps to isolate, identify and characterize urine particles on the screen to virtually eliminate the need for manual microscopy.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Urine Auto Analyzer
For Use With (Application) Urinalysis