Learn More
Beckman Coulter™ DXU 850M Iris
Catalog No. 01361048 Shop All Beckman Coulter Products
Change view
Click to view available options
DXU 850M Iris
Catalog No.
01-361-048 DXU 850M Iris
1 options
Catalog No. 01-361-048 Supplier Beckman Coulter™ Supplier No. C81579
Take the interruptions out of your urinalysis workflow.
Take the Interruptions Out of Your Urinalysis Workflow
The DxU Iris Workcell pairs the DxU 840/850 Iris urine microscopy analyzer with the AUTION MAX™ 4030 urine chemistry analyzer to create a scalable, fully automated urinalysis solution for your laboratory. Using proprietary Digital Flow Morphology technology with Auto-Particle Recognition (APR) software, the DxU Iris Workcell helps to isolate, identify and characterize urine particles on the screen to virtually eliminate the need for manual microscopy.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Urine Auto Analyzer
For Use With (Application) Urinalysis