Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DY 131, Thermo Scientific™

Catalog No. AAJ65171MA
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65171MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ65171MA Supplier Thermo Scientific Chemicals Supplier No. J65171MA

A selective agonist at estrogen-related receptors ERR→ and ERR→

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95167-41-2
Molecular Formula C18H21N3O2
Molecular Weight (g/mol) 311.385
MDL Number MFCD00715585
InChI Key WLKOCYWYAWBGKY-CPNJWEJPSA-N
Synonym unii-5vwv92039e, n-4-diethylaminobenzylidenyl-n'-4-hydroxybenzoyl-hydrazine, d06jpq, n'-4-diethylamino phenyl methylidene-4-hydroxybenzohydrazide, n-e-4-diethylamino phenyl methylideneamino-4-hydroxybenzamide, benzoic acid, p-hydroxy-, p-diethylamino benzylidene hydrazide, n'-1e-4-diethylamino phenyl methylene-4-hydroxybenzohydrazide, benzoic acid, 4-hydroxy-, 4-diethylamino phenyl methylene hydrazide, benzoic acid, 4-hydroxy-, 2-4-diethylamino phenyl methylene hydrazide, dy hplc
PubChem CID 5497124
IUPAC Name N-[(E)-[4-(diethylamino)phenyl]methylideneamino]-4-hydroxybenzamide
SMILES CCN(CC)C1=CC=C(C=C1)C=NNC(=O)C2=CC=C(C=C2)O

Specifications

Color Yellow
Quantity 10 mg
Formula Weight 311.38
Physical Form Solid
Chemical Name or Material DY 131
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.