Learn More
Apexbio Technology LLC DY131 95167-41-2 10mg
SDP
Catalog No. 5000095704
Encompass_Preferred
DY131 (95167-41-2) is a small-molecule agonist targeting the estrogen-related receptors ERR and ERR It is designed to selectively activate these orphan nuclear receptors thereby regulating estrogen-independent transcriptional pathways DY131 exerts its biological activity primarily through receptor-mediated activation of ERR and preferentially ERR with minimal effect on ERR In cell-based assays DY131 demonstrates moderate to strong agonist activity toward ERR / at low micromolar concentrations Based on these pharmacological properties DY131 holds research potential in investigating ERR-mediated transcriptional responses receptor selectivity and gene regulation particularly in prostate epithelial biology and oncology studies
Catalog No. 50-000-95704 Supplier Apexbio Technology LLC Supplier No. A337510
DY131 (95167-41-2) is a small-molecule agonist targeting the estrogen-related receptors ERR and ERR It is designed to selectively activate these orphan nuclear receptors thereby regulating estrogen-independent transcriptional pathways DY131 exerts its biological activity primarily through receptor-mediated activation of ERR and preferentially ERR with minimal effect on ERR In cell-based assays DY131 demonstrates moderate to strong agonist activity toward ERR / at low micromolar concentrations Based on these pharmacological properties DY131 holds research potential in investigating ERR-mediated transcriptional responses receptor selectivity and gene regulation particularly in prostate epithelial biology and oncology studies