Learn More
Medchemexpress LLC Dy131 (GSK 9089) | 95167-41-2 | C18H21N3O2 | 100 MG
SDP
Catalog No. 5000376063
Encompass_Preferred
DY131 is a potent and selective agonist for ERRγ and ERRβ. It has been observed to be inactive against ERRα, ERα, and ERβ. Additionally, it demonstrates inhibitory effects on Smo signaling. This product is intended for research purposes only and is not for patient use.
  • Potent and selective ERRγ and ERRβ agonist
  • Inactive against ERRα, ERα, and ERβ
  • Inhibits Smo signaling
  • For research use only
Catalog No. 50-003-76063 Supplier Medchemexpress LLC Supplier No. HY15483100MG
DY131 is a potent and selective agonist for ERRγ and ERRβ. It has been observed to be inactive against ERRα, ERα, and ERβ. Additionally, it demonstrates inhibitory effects on Smo signaling. This product is intended for research purposes only and is not for patient use.
  • Potent and selective ERRγ and ERRβ agonist
  • Inactive against ERRα, ERα, and ERβ
  • Inhibits Smo signaling
  • For research use only