Learn More
Medchemexpress LLC Dy131 (GSK 9089) | 95167-41-2 | C18H21N3O2 | 5 MG
SDP
Catalog No. 5000376923
Encompass_Preferred
DY131 (GSK 9089) is a potent and selective ERRγ and ERRβ agonist. It is inactive against ERRα, ERα, and ERβ, and also inhibits Smo signaling.
  • Potent and selective ERRγ and ERRβ agonist
  • Inactive against ERRα, ERα, and ERβ
  • Inhibits smo signaling
  • Suppresses cell proliferation in LNCaP-ERRγ and LNCaP cells
  • Inhibits Shh induced accumulation of Smo::EGFP
  • Decreases phosphorylated histone H3 (pH3) marked proliferation of CGNPs
Catalog No. 50-003-76923 Supplier Medchemexpress LLC Supplier No. HY154835MG
DY131 (GSK 9089) is a potent and selective ERRγ and ERRβ agonist. It is inactive against ERRα, ERα, and ERβ, and also inhibits Smo signaling.
  • Potent and selective ERRγ and ERRβ agonist
  • Inactive against ERRα, ERα, and ERβ
  • Inhibits smo signaling
  • Suppresses cell proliferation in LNCaP-ERRγ and LNCaP cells
  • Inhibits Shh induced accumulation of Smo::EGFP
  • Decreases phosphorylated histone H3 (pH3) marked proliferation of CGNPs