Learn More
Medchemexpress LLC DYB-03 | 2982792-85-6 | 99.5% | 479.56 | 1 ML
SDP
Catalog No. 5000395312
Encompass_Preferred
DYB-03 is an orally active HIF-1α/EZH2 inhibitor. It inhibits the migration, invasion, and angiogenesis of lung cancer cells and HUVECs both in vitro and in vivo. Additionally, DYB-03 induces apoptosis in A549 and H460 cells that are resistant to 2-ME2 and GSK126.
  • Oral active HIF-1α/EZH2 inhibitor
  • Inhibits migration, invasion, and angiogenesis of lung cancer cells and HUVECs (in vitro and in vivo)
  • Induces apoptosis in 2-ME2- and GSK126-resistant A549 and H460 cells
Catalog No. 50-003-95312 Supplier Medchemexpress LLC Supplier No. HY16131310MM/1ML
DYB-03 is an orally active HIF-1α/EZH2 inhibitor. It inhibits the migration, invasion, and angiogenesis of lung cancer cells and HUVECs both in vitro and in vivo. Additionally, DYB-03 induces apoptosis in A549 and H460 cells that are resistant to 2-ME2 and GSK126.
  • Oral active HIF-1α/EZH2 inhibitor
  • Inhibits migration, invasion, and angiogenesis of lung cancer cells and HUVECs (in vitro and in vivo)
  • Induces apoptosis in 2-ME2- and GSK126-resistant A549 and H460 cells