Learn More
DYCEM INC Dycem Dyclean Concentrate
Catalog No. NC0513891
Encompass_Preferred
Dyclean is an all-purpose, water soluable concentrated cleaning solution for regular cleaning in most applications outside the cleanroom enviroment. Use diluted 1:20 with water for regular maintenance. It can be used for heavy duty cleaning as needed. This product is recommended for cleaning Dycem mats OUTSIDE of the cleanroom to maintain particle removing qualities.
Catalog No. NC0513891 Supplier DYCEM INC Supplier No. CZCLDCGUS
Dyclean is an all-purpose, water soluable concentrated cleaning solution for regular cleaning in most applications outside the cleanroom enviroment. Use diluted 1:20 with water for regular maintenance. It can be used for heavy duty cleaning as needed. This product is recommended for cleaning Dycem mats OUTSIDE of the cleanroom to maintain particle removing qualities.