Learn More
Apexbio Technology LLC Dyclonine HCl 536-43-6 5g
SDP
Catalog No. 5000090016
Encompass_Preferred
Dyclonine HCl (CAS 536-43-6) is a small-molecule inhibitor targeting voltage-gated sodium channels It is designed to reversibly block neuronal sodium channel activity thereby inhibiting the propagation of nerve impulses and regulating pain signaling pathways Dyclonine HCl exerts its biological activity primarily through blockade of voltage-gated sodium channels on neuronal membranes In in vitro studies dyclonine HCl demonstrates inhibitory activity with an IC50 value typically reported in the range of 150 300 M depending on experimental conditions Based on these pharmacological properties dyclonine HCl holds research potential in studies of sodium channel activity neuronal excitability and pain signaling
Catalog No. 50-000-90016 Supplier Apexbio Technology LLC Supplier No. B19365000
Dyclonine HCl (CAS 536-43-6) is a small-molecule inhibitor targeting voltage-gated sodium channels It is designed to reversibly block neuronal sodium channel activity thereby inhibiting the propagation of nerve impulses and regulating pain signaling pathways Dyclonine HCl exerts its biological activity primarily through blockade of voltage-gated sodium channels on neuronal membranes In in vitro studies dyclonine HCl demonstrates inhibitory activity with an IC50 value typically reported in the range of 150 300 M depending on experimental conditions Based on these pharmacological properties dyclonine HCl holds research potential in studies of sodium channel activity neuronal excitability and pain signaling