Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dyclonine hydrochloride | 536-43-6 | 99.3% | 1 ML
SDP

Catalog No. 5000406272
Encompass_Preferred

Dyclonine hydrochloride, also known as Dyclocaine hydrochloride, is an orally effective covalent inhibitor of Aldehyde Dehydrogenase (ALDH) that can cross the blood-brain barrier. It demonstrates sensitizing activities for targeted cancer cells and exhibits antibacterial properties. Additionally, Dyclonine hydrochloride functions as a local anesthetic, capable of suppressing or relieving pain by blocking the transmission of nerve impulses and inhibiting the sensation of touch and pain.

  • Orally effective ALDH covalent inhibitor
  • Crosses the blood-brain barrier
  • IC50 of 35 μM for ALDH2 and 76 μM for ALDH3A1
  • Sensitizes targeted cancer cells
  • Possesses antibacterial activity
  • Acts as a local anesthetic to suppress and relieve pain
  • Blocks the transmission of nerve impulses
  • Inhibits the sensation of touch and pain

Catalog No. 50-004-06272 Supplier Medchemexpress LLC Supplier No. HYB0364A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dyclonine hydrochloride, also known as Dyclocaine hydrochloride, is an orally effective covalent inhibitor of Aldehyde Dehydrogenase (ALDH) that can cross the blood-brain barrier. It demonstrates sensitizing activities for targeted cancer cells and exhibits antibacterial properties. Additionally, Dyclonine hydrochloride functions as a local anesthetic, capable of suppressing or relieving pain by blocking the transmission of nerve impulses and inhibiting the sensation of touch and pain.

  • Orally effective ALDH covalent inhibitor
  • Crosses the blood-brain barrier
  • IC50 of 35 μM for ALDH2 and 76 μM for ALDH3A1
  • Sensitizes targeted cancer cells
  • Possesses antibacterial activity
  • Acts as a local anesthetic to suppress and relieve pain
  • Blocks the transmission of nerve impulses
  • Inhibits the sensation of touch and pain

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.