Learn More
Medchemexpress LLC Dyclonine hydrochloride | 536-43-6 | 99.3% | 10 G
SDP
Catalog No. 5000406613
Encompass_Preferred
Dyclonine hydrochloride is an orally effective ALDH covalent inhibitor that crosses the blood-brain barrier. It functions as a local anesthetic, capable of suppressing or relieving pain by blocking nerve impulse transmission. It also exhibits sensitizing activities for targeted cancer cells and possesses antibacterial and fungicidal properties, demonstrating significant activity against various bacteria and fungi.
  • Orally effective ALDH covalent inhibitor
  • Crosses the blood-brain barrier
  • Functions as a local anesthetic
  • Sensitizes cancer cells to cysteine and GSH deficiency
  • Demonstrates bactericidal and fungicidal activity
Catalog No. 50-004-06613 Supplier Medchemexpress LLC Supplier No. HYB0364A10G
Dyclonine hydrochloride is an orally effective ALDH covalent inhibitor that crosses the blood-brain barrier. It functions as a local anesthetic, capable of suppressing or relieving pain by blocking nerve impulse transmission. It also exhibits sensitizing activities for targeted cancer cells and possesses antibacterial and fungicidal properties, demonstrating significant activity against various bacteria and fungi.
  • Orally effective ALDH covalent inhibitor
  • Crosses the blood-brain barrier
  • Functions as a local anesthetic
  • Sensitizes cancer cells to cysteine and GSH deficiency
  • Demonstrates bactericidal and fungicidal activity