Learn More
Medchemexpress LLC Dyclonine hydrochloride | 536-43-6 | 99.3% | 5 G
SDP
Catalog No. 5000406273
Encompass_Preferred
Dyclonine hydrochloride is an orally effective ALDH covalent inhibitor that crosses the blood-brain barrier. It has an IC50 of 35 μM for ALDH2 and 76 μM for ALDH3A1. This compound exhibits sensitizing activities for targeted cancer cells and possesses antibacterial properties. Additionally, it functions as a local agent, capable of suppressing or relieving pain by blocking nerve impulse transmission and inhibiting the sensation of touch and pain.
  • Orally effective ALDH covalent inhibitor
  • Crosses the blood-brain barrier
  • Sensitizes targeted cancer cells
  • Exhibits antibacterial properties
  • Suppresses or relieves pain as a local agent
  • Blocks transmission of nerve impulses
  • Inhibits sensation of touch and pain
Catalog No. 50-004-06273 Supplier Medchemexpress LLC Supplier No. HYB0364A5G
Dyclonine hydrochloride is an orally effective ALDH covalent inhibitor that crosses the blood-brain barrier. It has an IC50 of 35 μM for ALDH2 and 76 μM for ALDH3A1. This compound exhibits sensitizing activities for targeted cancer cells and possesses antibacterial properties. Additionally, it functions as a local agent, capable of suppressing or relieving pain by blocking nerve impulse transmission and inhibiting the sensation of touch and pain.
  • Orally effective ALDH covalent inhibitor
  • Crosses the blood-brain barrier
  • Sensitizes targeted cancer cells
  • Exhibits antibacterial properties
  • Suppresses or relieves pain as a local agent
  • Blocks transmission of nerve impulses
  • Inhibits sensation of touch and pain