Learn More
Medchemexpress LLC Dydrogesterone-d6 | 152-62-5 | 1 MG
SDP
Catalog No. 5000405952
Encompass_Preferred
Dydrogesterone-d6 is a deuterium-labeled version of Dydrogesterone. Dydrogesterone is an orally active and potent progestogen, used for various gynecological conditions resulting from progesterone deficiency. It is intended for research use only and not for patient use. Deuterium substitution in drug molecules is being explored for its potential to influence pharmacokinetic and metabolic profiles.
  • Can be utilized as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-05952 Supplier Medchemexpress LLC Supplier No. HYB0257AS1MG
Dydrogesterone-d6 is a deuterium-labeled version of Dydrogesterone. Dydrogesterone is an orally active and potent progestogen, used for various gynecological conditions resulting from progesterone deficiency. It is intended for research use only and not for patient use. Deuterium substitution in drug molecules is being explored for its potential to influence pharmacokinetic and metabolic profiles.
  • Can be utilized as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.