Learn More
AMPLYUS LLC miniPCR bio Dye Electrophoresis Lab: Molecular Rainbow
SDP
Catalog No. NC2113643
Encompass_Preferred
Dye Electrophoresis Lab: Molecular Rainbow. Separate molecular mixtures and learn principles of gel electrophoresis using safe dyes that simulate DNA — no visualization equipment required.
This item is not returnable. View return policy
Catalog No. NC2113643 Supplier AMPLYUS LLC Supplier No. KT140001
Dye Electrophoresis Lab: Molecular Rainbow. Separate molecular mixtures and learn principles of gel electrophoresis using safe dyes that simulate DNA — no visualization equipment required.