Learn More
Bachem Tide FluorTM 2-LL-37 Trifluoroacetate
SDP
Supplier:  Bachem 4104170.0100
Encompass_Preferred
Tide FluorTM 2-LL-37 Trifluoroacetate, Tide FluorTM 2-Leu-Leu-Gly-Asp-Phe-Phe-Arg-Lys-Ser-Lys-Glu-Lys-Ile-Gly-Lys-Glu-Phe-Lys-Arg-Ile-Val-Gln-Arg-Ile-Lys-Asp-Phe-Leu-Arg-Asn-Leu-Val-Pro-Arg-Thr-Glu-Ser-OH, , C205H340N60O53 + dye, Mr 4962, 0.1mg. Dye-labeled LL-37, excitation 500nm, emission 527nm. Tide FluorTM-2 shows an excellent photostability and brightness, its fluorescence is pH-insensitive.
Catalog No. 50-194-6669