Learn More
DC Scientific Glass Inc DYED GEL, 40 GR
SDP
Catalog No. NC1656599
Encompass
DYED GEL, 40 GRAM BOTTLE (FOR GASOLINE ONLY, NOT FOR USE WITH DIESEL OR JET FUEL)
Catalog No. NC1656599 Supplier DC Scientific Glass Inc Supplier No. DCP1319009
DYED GEL, 40 GRAM BOTTLE (FOR GASOLINE ONLY, NOT FOR USE WITH DIESEL OR JET FUEL)