Learn More
Biotium Dyes For Labeling Carbonyls and Carbohydrates Fluorescence Microscopy Cell and Organelle Stains Cell Surface and Membrane Dyes 1/EA
Catalog No. 5000501403
Encompass_Preferred
Label live cell surface glycoproteins with visible red fluorescent dye (Ex/Em 550/570 nm) for fixable plasma membrane staining GlycoLiner uses novel reactive chemistry for faster more efficient and gentler labeling compared to conventional methods 500 reactions for imaging (200 uL volume)
Catalog No. 50-005-01403 Supplier Biotium Supplier No. 30151
Label live cell surface glycoproteins with visible red fluorescent dye (Ex/Em 550/570 nm) for fixable plasma membrane staining GlycoLiner uses novel reactive chemistry for faster more efficient and gentler labeling compared to conventional methods 500 reactions for imaging (200 uL volume)