Learn More
Biotium Dyes For Labeling Carbonyls and Carbohydrates Reactive CF Dyes Other Dyes Biotinylation Reagents Reactive CF Dyes CF405 1/EA
Catalog No. 5000498436
Encompass_Preferred
Aminooxy dyes have an aminooxy reactive group which can be used to label a target molecule with an aldehyde or ketone group An aminooxy group readily reacts with an aldehyde or ketone group to form a stable linkage without the use of a reducing agent CF dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes Blue fluorescent CF405M dye has excitation/emission at 408/452 nm
Catalog No. 50-004-98436 Supplier Biotium Supplier No. 92055
Aminooxy dyes have an aminooxy reactive group which can be used to label a target molecule with an aldehyde or ketone group An aminooxy group readily reacts with an aldehyde or ketone group to form a stable linkage without the use of a reducing agent CF dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes Blue fluorescent CF405M dye has excitation/emission at 408/452 nm