Learn More
Proteintech Group Inc DYKDDDDK Fab-Trap™ Agarose Kit
SDP
Supplier:  Proteintech Group Inc FFAK20
Encompass
The ChromoTek DYKDDDDK Fab-Trap™ Agarose Kit consists of an anti-DYKDDDDK-tag Fab fragment, which is covalently bound to agarose beads. DYKDDDDK Fab-Trap™ Agarose Kit is used to immunoprecipitate 1x and 3xDYKDDDDK-tagged fusion proteins from cell extracts of various organisms like mammals, plants, bacteria, yeast, insects etc
Catalog No. 50-225-8211