Learn More
Cayman Chemical DYKDDDDK-HRP Tracer 1mL
SDP
Catalog No. 502943419
Encompass_Preferred
DYKDDDDK-HRP Tracer has been tested and formulated to work exclusively with Cayman’s DYKDDDDK-Tag Detection ELISA Kit (Item No. 501560).
Catalog No. 50-294-3419 Supplier Cayman Chemical Supplier No. 4015961 ML
DYKDDDDK-HRP Tracer has been tested and formulated to work exclusively with Cayman’s DYKDDDDK-Tag Detection ELISA Kit (Item No. 501560).