Learn More
Cayman Chemical DYKDDDDK Peptide (trifluoroacetate salt), Purity: ≥98%, 10 mg
SDP
Catalog No. 502947581
Encompass_Preferred
A octapeptide fusion tag; has commonly been used as an epitope tag on recombinant proteins to facilitate their detection and purification by DYKDDDDK-specific antibodies in vitro and in vivo
Catalog No. 50-294-7581 Supplier Cayman Chemical Supplier No. 3319810 MG
A octapeptide fusion tag; has commonly been used as an epitope tag on recombinant proteins to facilitate their detection and purification by DYKDDDDK-specific antibodies in vitro and in vivo