Learn More
Cayman Chemical DYKDDDDK Polyclonal Antibody, 1 ea
SDP
Catalog No. 502971840
Encompass_Preferred
Immunogen: DYKDDDDK peptide sequence • Host: Rabbit • Application: (+) WB; (−) IP
Catalog No. 50-297-1840 Supplier Cayman Chemical Supplier No. 1621501 EA
Immunogen: DYKDDDDK peptide sequence • Host: Rabbit • Application: (+) WB; (−) IP