Learn More
MSC Dykem High Temp Marker, White, Medium Tip, 2000, 1 ea
Catalog No. NC1692864
Encompass
The high temperature marker is specifically formulated for marking surfaces exposed to extreme high-temperature environments. Marks are applied at room temperature to almost any non-porous surface, including metal, glass, and ceramic. When used on ceramics, the marks become permanently fused after heat is applied.
  • Fiber tip paint marker with valve-action
  • Valve-action mechanism prevents paint in the barrel from drying if the cap is left off
  • Threaded barrel allows the marker to be screwed into a jig or apparatus
  • Withstands temperatures up to 2000°F (1093°C)
  • Suitable for marking metal, glass, and ceramic
  • Drying time of 45-60 seconds
  • Medium, felt tip
Catalog No. NC1692864 Supplier MSC Supplier No. 04400529
The high temperature marker is specifically formulated for marking surfaces exposed to extreme high-temperature environments. Marks are applied at room temperature to almost any non-porous surface, including metal, glass, and ceramic. When used on ceramics, the marks become permanently fused after heat is applied.
  • Fiber tip paint marker with valve-action
  • Valve-action mechanism prevents paint in the barrel from drying if the cap is left off
  • Threaded barrel allows the marker to be screwed into a jig or apparatus
  • Withstands temperatures up to 2000°F (1093°C)
  • Suitable for marking metal, glass, and ceramic
  • Drying time of 45-60 seconds
  • Medium, felt tip